Explosion-Proof Blowers for Explosion-Proof Laboratory Hoods, Labconco®
Supplier: Labconco
7068100 7068300 7068500 7068700 7068900 7069100 7069300 7069500 7069700 7180100 7180300 7180500 7180700 7180900 7181100 7181300 7181500 7181700 7182100 7182300
82006-758EA 6256.8 CAD
82006-758 82006-761 82006-762 82006-764 82006-766 82006-768 82006-770 82006-772 82006-774 82006-798 82006-800 82006-802 82006-804 82006-806 82006-808 82006-810 82006-812 82006-814 82006-816 82006-818
Explosion-Proof Blowers for Explosion-Proof Laboratory Hoods, Labconco®
Air Purifiers
Fiberglass explosion-proof blowers are designed for fume hood exhaust systems in moderate to highly corrosive conditions and have non-sparking polypropylene impellers and explosion-proof motors.
Coated steel explosion-proof blowers are recommended for low to moderately corrosive applications and feature non-sparking aluminum impellers and explosion-proof motors for maximum safety. Housing has phenolic coating for corrosion resistance.
Both blowers have a dry powder epoxy-coated base and weather cover for protection in extreme weather. Sheaves are adjustable to allow for air volumeMore Product Information