Cleanroom Labels, CleanMark®, LG INTERNATIONAL
Supplier: Lgi
L10018 L10020 L46040 L65282
82017-982RL 444.45 CAD
82017-982 82006-730 82006-731 89204-788 10799-268
Cleanroom Labels, CleanMark®, LG INTERNATIONAL
Labels
Cleanroom Labels recommended for use in any cleanroom environment.
  • Labels can be printed upon with a cleanroom pen or by any standard thermal transfer printer
  • Manufactured from 100% synthetic
  • Nonparticulating
  • Ultra low-outgassing raw materials
These removable labels can be easily removed without leaving behind adhesive residue contamination.
CleanMark® labels can be custom constructed to fit any size, and can be used with either direct thermal or thermal transfer printers. These labels can withstand extreme hot or cold temperatures, harsh chemicalMore Product Information