Potassium nitrate (nitre- KNO₃)

Supplier: Avantor
Warning

80705-020EA 323.95 CAD
80705-020
Potassium nitrate (nitre- KNO₃)
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR