VWR® Signature™ Coveralls made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR International
89012-828 89012-830 CA89012-824 CA89012-826 CA89012-832 CA89012-834 CA89012-822 CA89012-828 CA89012-830 89012-822 89012-824 89012-826 89012-832 80200-620 80200-622 80200-624 80200-626 80200-628 89012-834 89127-188 89128-778 89128-780 89128-782 89128-784
89012-828CS 975.12 CAD
89012-828 89012-830 CA89012-824 CA89012-826 CA89012-832 CA89012-834 CA89012-822 CA89012-828 CA89012-830 89012-822 89012-824 89012-826 89012-832 80200-620 80200-622 80200-624 80200-626 80200-628 89012-834 89127-188 89128-778 89128-780 89128-782 89128-784
VWR® Signature™ Coveralls made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Protective, durable, and comfortable coveralls provide an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
  • Protective, durable, and comfortable
  • Excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
  • Chemically and biologically inert, low linting, and static free until laundered
  • Serged seams, laydown collar, front zipper closure, and elastic wrists
  • Available with elastic ankles, with attached Gripper™ boots, or with attached boots and hood
  • Boot attachments combine embossed PVC exterior sole
More Product Information