HyFlex® 11-100 Static Control Gloves, Ansell
Supplier: Ansell Healthcare
205593
80092-584CS 3867.54 CAD
80092-584
HyFlex® 11-100 Static Control Gloves, Ansell
Gloves
Designed for micro-electronic work where electro-static discharge (ESD) must be avoided, these comfortable industrial gloves with nitrile foam ensure that surfaces stay clean and uncontaminated. X-Static® yarn and static-reducing coating help prevent damage to sensitive electronic components and injury.
  • Ensure uncontaminated surfaces
  • Static-reducing coating
  • Oil is channeled away for a sure grip
  • Tolerance: ±0.5"
A thin design improves the handling of ever smaller components, while the ergonomic liner allows for precision in assembly applications.