SP Wilmad-LabGlass NMR Microcell Assemblies, 5 mm, SP Industries
Supplier: SP Industries
529-A 529-D 529-E 529-B 529-C
80087-498PK 189.52 CAD
80087-498 80087-500 80087-502 80087-504 80087-506 80087-508 80087-510
SP Wilmad-LabGlass NMR Microcell Assemblies, 5 mm, SP Industries
Tubes NMR Tubes
Microcell assemblies are designed to facilitate the spectra analysis of small samples in standard 5 mm thin-wall NMR tubes
Note: Not recommended for variable temperature applications because of differences in coefficients of thermal expansion between materials. The precision nature of the match is only acceptable at room temperature.
Assemblies consist of an insert, a PTFE holder, and a positioning rod; all components are sold separately. To suit your sample size and requirements, choose either spherical bulb, capillary tube, or cylindrical bulb. The PTFE holder holds the insert in theMore Product Information