Conductivity Cells, WTW
Supplier: WTW
203730 301602 301961 301962 301970 301971 301973 301960
80085-444EA 1925.77 CAD
80085-444 80085-574 80085-578 80085-580 80085-582 80085-584 80085-586 97042-964 80085-577
Conductivity Cells, WTW
Electrodes Conductivity Sensors
This selection of conductivity cells includes the TetraCon® series of cells, which feature the highest degree of precision and linearity by optimized cell geometry, an extremely large measuring range with just one cell, long-term cell constant stability with high-quality abrasion-resistant graphite electrodes, and a built-in temperature sensor
The flow-thru vessel is constructed of transparent PVC and has a 18mm I.D. Filling volume (without sensor): 13mL.
TetraCon® cells also feature the smallest immersion depth possible and have no measuring errors due to cable influences, primary orMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable