SenTix® pH Electrodes, WTW
Supplier: WTW
103630 103631 103633 103635 103636 103637 103639 103640 103641 103642 103643 103645 103647 103650 103646 103651 103652 103660 103661 103695
80085-210EA 807.19 CAD
80085-210 80085-212 80085-214 80085-216 80085-218 80085-220 80085-222 80085-224 80085-226 80085-228 80085-230 80085-232 80085-234 80085-240 97042-792 97042-794 97042-796 97042-798 97042-800 97042-802 97042-806 97042-814 97042-816 97042-818 97042-820 97042-822 97042-824 97042-826 97042-830
SenTix® pH Electrodes, WTW
Electrodes pH Electrodes
Low-resistance glass membranes guarantee stable measuring signals even at low temperatures
Low-maintenance pH electrodes with gel electrolyte are ideal for portable measurements, as well as for routine measurements in-the-laboratory. With or without built-in temperature probe all electrodes have robust plastic shafts with a low-maintenance gel reference system.
Special electrodes for unique applications need special electrodes. SenTix special electrodes can take on the challenges associated with measuring the pH value of surfaces, solids, suspensions, emulsions, low ionic samples,More Product Information