Crews® Class II Traffic Safety Vests, MCR Safety
Supplier: MCR Safety
CL2MLL CL2MLX2 CL2MLX3 CL2MLX4 CL2MLXL CL2MOL CL2MOX2 CL2MOX3 CL2MOX4 CL2MOXL
80082-396EA 43.43 CAD
80082-396 80082-398 80082-400 80082-402 80082-404 80082-406 80082-408 80082-410 80082-412 80082-414
Crews® Class II Traffic Safety Vests, MCR Safety
Workwear
Class II safety vests are designed for use in occupational activities that require greater worker visibility due to inclement weather, complex backgrounds, or proximity to vehicular traffic exceeding 25mph
  • 100% polyester tear-away mesh vests are available in two fluorescent colors
  • One horizontal 5cm (2") silver reflective strip and two vertical 5cm (2") silver reflective strips
  • Velcro® closures on the front, both shoulders, and garment sides allow for quick and easy donning and removal
These vests are ideal for use in utility, survey, railway,More Product Information