PVC/Polyester Garment, Detachable Hood, Snap Front, MCR Safety
Supplier: MCR Safety
2003L 2003M 2003S 2003X2 2003X3 2003X4 2003X5 2003X7 2003XL
80082-254EA 51.07 CAD
80082-254 80082-256 80082-258 80082-260 80082-262 80082-264 80082-266 80082-270 80082-272
PVC/Polyester Garment, Detachable Hood, Snap Front, MCR Safety
Workwear
The Classic Series Material is the most popular in the Industry
  • Jacket features a snap front, take-up snaps at the wrists, detachable hood with drawstring, and two front patch pockets
  • Bib pants feature a fly front with snap and take-up snaps at the waist and ankles
It combines the strength and waterproof qualities of PVC with the durability of a polyester backing. The 2003 yellow three piece suit features a jacket with detachable hood and bib style pants.
Order Now
SPECIFICATIONS
Environmentally Preferable
Garment application Industrial,Laboratory
Risk area
Gender
Sleeve or leg length Long
Closure type Press studs / snaps
Fabric PVC/polyester
Thickness 0.35 mm