Plasticized Cuff Gloves, Select Grade, MCR Safety
Supplier: MCR Safety
1400S 1400XXL
80081-246PK 355.39 CAD
80081-246 80081-250
Plasticized Cuff Gloves, Select Grade, MCR Safety
Gloves
Select shoulder gloves provide you with excellent protection in harsh working environments.
  • Shirred elastic back
  • Leather knuckle strap rounds off the features of this rugged offering
The 1400 leather palm glove features A grade select shoulder cow leather with a launderable white canvas back. It also features a fleecy lining in the palm and a 15 ounce 2.5" plasticized safety cuff.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Liner material Fleece
Cuff style Safety cuff