Dry Block Heater, Digital, 240 V Glas-Col®
Supplier: Glas-Col
099AHBC24 R34 562 R34 561 R34563 R34516
80078-740EA 3518.27 CAD
80078-740 80078-742 80078-744 80078-746 80078-748
Dry Block Heater, Digital, 240 V Glas-Col®
Block Heaters Digital Block Heaters
This small, constant-temperature heater can accommodate a variety of different size blocks
  • 240 V
Maintains temperature from slightly above ambient to 125°C (257°F), with ±1°C accuracy. The heater opening is in standard SBS format, which allows heating of high- or low-profile standard well plates. The heater features single-loop, auto-tuning digital control with simple input commands, dual four-digit LED display for temperature in °C or °F, and up and down keys for set point adjustment. The heater also features PID control parameters that setMore Product Information