Stopcocks, Straight Bore, Glass Plugs, Capillary Arms, Chemglass
Supplier: Chemglass
CG-406-02
80074-076EA 136.35 CAD
80074-076
Stopcocks, Straight Bore, Glass Plugs, Capillary Arms, Chemglass
Stopcocks
Similar to the CG-400 series, but with the sidearms being made of capillary tubing having the same bore as the plug.
Delivery information: Supplied complete with metal retaining clip CG-423.