Funnels, Addition, Graduated, Threaded Valves, Chemglass
Supplier: Chemglass
CG-1714-06 CG-1714-01 CG-1714-02 CG-1714-03 CG-1714-04 CG-1714-10 CG-1714-12 CG-1714-16
80070-734EA 1145.16 CAD
80070-734 80070-724 80070-726 80070-728 80070-730 80070-738 80070-742 80070-750
Funnels, Addition, Graduated, Threaded Valves, Chemglass
Funnels Dropping Funnels
Cylindrical style graduated addition funnel has a threaded, grease-free extended tip valve.
  • Design of all PTFE plug permits semi-needle valve action
  • With a standard taper top outer joint and lower inner joint
  • Both joints being the same size, and pressure equalizing arm
Delivery information: Funnels with 14/20 and 19/22 joints are supplied complete with a stopper.