Round-Bottom Boiling Flasks with Three Angled Necks, Heavy Wall, Chemglass
Supplier: Chemglass
CG-1524-08 CG-1524-12 CG-1524-14 CG-1524-15 CG-1524-16 CG-1524-17 CG-1524-19 CG-1524-20 CG-1524-21 CG-1524-22 CG-1524-26 CG-1524-31 CG-1524-33 CG-1524-35 CG-1524-36 CG-1524-46 CG-1524-48 CG-1524-50 CG-1524-51 CG-1524-53 CG-1524-55 CG-1524-59 CG-1524-60 CG-1524-62 CG-1524-67 CG-1524-68 CG-1524-83 CG-1524-A-01 CG-1524-A-09 CG-1524-A-10 CG-1524-A-15 CG-1524-T-01 CG-1524-T-03 CG-1524-T-05 CG-1524-01 CG-1524-02 CG-1524-03 CG-1524-05 CG-1524-07 CG-1524-37 CG-1524-38 CG-1524-40 CG-1524-41 CG-1524-42 CG-1524-82 CG-1524-90 CG-1524-A-02 CG-1524-A-03 CG-1524-A-05 CG-1524-A-06 CG-1524-A-08 CG-1524-A-11 CG-1524-A-12 CG-1524-A-13 CG-1524-A-14 CG-1524-A-16 CG-1524-A-04 CG-1524-A-07
80069-458EA 562.08 CAD
80069-458 80069-460 80069-464 80069-466 80069-468 80069-470 80069-474 80069-476 80069-478 80069-480 80069-488 80069-498 80069-500 80069-504 80069-506 80069-522 80069-526 80069-530 80069-532 80069-536 80069-540 80069-548 80069-550 80069-554 80069-564 80069-566 80069-596 80069-610 80069-626 80069-628 80069-638 89028-598 89028-600 89028-602 80069-448 80069-450 80069-452 80069-454 80069-456 80069-508 80069-510 80069-512 80069-514 80069-516 80069-594 80069-602 80069-612 80069-614 80069-618 80069-620 80069-624 80069-630 80069-632 80069-634 80069-636 80069-640 10170-850 10170-852
Round-Bottom Boiling Flasks with Three Angled Necks, Heavy Wall, Chemglass
Flasks Round Bottom Flasks
These round-bottom, heavy wall, three-neck flasks have [ST] outer joints on the center neck and 20°-angled side necks
Both side necks and center neck have the same joint size.