Airfree® Schlenk Double Bubbler, Chemglass
Supplier: Chemglass
AF-0513-D-21
80064-568EA 304.5 CAD
80064-568
Airfree® Schlenk Double Bubbler, Chemglass
Bubblers
Double bubbler with two oil reservoirs.
  • Used for monitoring gas flow to reactions while providing a vent to the atmosphere
  • Reservoir head prevents oil from being sucked back into the system
Bubbler is approximately 125 W×135 mm H. Hose connections are 9 mm O.D. at the middle serration.
Order Now
SPECIFICATIONS
Environmentally Preferable