Airfree® Bubbler, Chemglass
Supplier: Chemglass
AF-0513-20
80064-558EA 156.41 CAD
80064-558
Airfree® Bubbler, Chemglass
Bubblers
Used to monitor gas flow to reactions while providing a vent to the atmosphere. Reservoir head prevents oil from being sucked back into the system.
Bubbler has an 8 mm O.D. top and side tubulations.