Round Bottom Flasks, Minum-ware, MW-58, Chemglass
Supplier: Chemglass
MW-58-02
80064-168EA 84.29 CAD
80064-168
Round Bottom Flasks, Minum-ware, MW-58, Chemglass
Flasks Round Bottom Flasks
Micro round bottom flask with a 14/10 threaded outer joint
Supplied complete with an open-top screw cap and Viton O-ring.