Stir Bar Kit, Egg-Shaped, PTFE, Chemglass
Supplier: Chemglass
CG-2005-20
80062-154EA 818.18 CAD
80062-154
Stir Bar Kit, Egg-Shaped, PTFE, Chemglass
Stirrers Magnetic Stirring Bars
Egg-shaped stir bars coated with PTFE
Kit contains six each of 20x10mm bars, six each of 30x16mm bars, three each of 40x20mm bars, two each of 50x20mm bars, and one 64x20mm bar. Replacement stir bars are available separately.