Glas-Col® Heating Tapes, Ribbon-Type Elements
Supplier: Chemglass
CG-14050-01 CG-14050-02 CG-14050-04 CG-14050-05 CG-14050-06 CG-14050-07 CG-14050-08 CG-14050-10 CG-14050-11 CG-14050-12 CG-14050-16 CG-14050-17 CG-14050-18 CG-14050-19
80061-072EA 369.34 CAD
80061-072 80061-074 80061-078 80061-080 80061-082 80061-084 80061-086 80061-088 80061-090 80061-092 80061-098 80061-100 80061-102 80061-104
Glas-Col® Heating Tapes, Ribbon-Type Elements
Tapes
GLAS-COL® Tape heats odd lengths of glass or metal pipe
Ribbon-type heating elements running lengthwise in the tape are encased in durable glass braid and are impregnated and covered with tough silicone rubber.
The silicone rubber insulates stranded terminal leads at one end and waterproofs the fittings at both ends. It also permits use in high-moisture areas and helps to make the tapes resistant to most laboratory environment conditions. All tapes are supplied with a 2 blade plug and have a temperature limit of 249 °C (480 °F).
Caution: Note: Power ratings listed are for tapes notMore Product Information