Rotary Evaporator Adapters, Vial, Chemglass
Supplier: Chemglass
CG-1318-22 CG-1318-40 CG-1318-20 CG-1318-21 CG-1318-23 CG-1318-24 CG-1318-25 CG-1318-30 CG-1318-10 CG-1318-12
80060-918EA 72.05 CAD
80060-918 89028-420 80060-914 80060-916 80060-920 80060-922 80060-924 80060-926 80068-042 80068-046
Rotary Evaporator Adapters, Vial, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapters permit the connection of standard laboratory dram vials directly to the vapor tube or bump trap of a rotary evaporator.
  • Adapters have a 20-400 GPI thread finish at the bottom
  • A fluoropolymer resin lined, polypropylene double-threaded connector attaches the vial to the adapter
Ordering information: The glass adapter and double-threaded connections must be ordered separately. Connector set includes one of each of the plastic connectors.
Order Now
SPECIFICATIONS
Environmentally Preferable