Glas-Col® Series M Spherical Heating Mantles, Chemglass
Supplier: Chemglass
CG-11000-07 CG-11000-08 CG-11000-09 CG-11000-10 CG-11000-11 CG-11000-12 CG-11000-13
80060-342EA 1090.42 CAD
80060-342 80060-344 80060-346 80060-348 80060-350 80060-352 80060-354
Glas-Col® Series M Spherical Heating Mantles, Chemglass
Heaters Heating Mantles
The Glas-Col® Series M mantle is the most compact mantle, with an aluminum shell that can heat flasks up to 72 liters.
  • Durable aluminum housing on the exterior and resilient glass fabric on the interior
  • The internal operating temperature is 450 °C
  • All mantles are 115 Volt, and are supplied with a detachable 4', 3-wire cord
Like the TM series, this mantle has a durable aluminum housing outside and resilient glass fabric inside to support flasks and protect flask walls from thermal strain. Only the bottom of the flask is covered, so the contents beingMore Product Information