EasyClean® 360 Knitted Polyester Padded Isolator Cleaning Tool Covers
Supplier: Berkshire
EC360CHP40 EC360PAD6
77609-762CS 1132.53 CAD
77609-762 78019-702
EasyClean® 360 Knitted Polyester Padded Isolator Cleaning Tool Covers
Mops
The knitted polyester padded cover and rounded edges of the mop head ensure the tool does not scratch or damage surfaces.
  • Made from Berkshire’s Choice® SuperSorb 100% continuous filament knitted polyester material
  • Laundered and packaged in an ISO Class 4 cleanroom for superior fiber, particle, and residue performance
  • Foam-free cushioned design ensures even surface coverage with reduced residues and contaminants
  • Easy installation and secure fit on the mop head
  • Recommended for use with the EasyClean® 360 Knitted Polyester Elastic Covers
  • Compatible for use in
More Product Information