Static Mouse Cage 1264C
Supplier: Tecniplast Usa
1264C025 1264C524 ACBT0402 ACBT0402SU 1264C016 1264C051 1264C400SUC
75876-744EA 78.97 CAD
75876-744 75876-746 75876-808 77969-762 77969-654 77969-656 77969-660
Static Mouse Cage 1264C
Animal Supplies Animal Cages
The 1264C mouse cage can comfortably house 3 mice of standard size and is available in polycarbonate or polysulfone materials.
  • Made from transparent plastic with exceptional impact strength and heat resistance
  • 57 square inch cage base
  • Compatible with a variety of lids, tops, accessories and racks
  • Stainless steel accessories
  • Can house 3 mice of standard size
The Tecniplast 1264C conventional cage is designed for housing laboratory rodents, particularly mice, offering a standard size and configuration that ensures compatibility with other equipment in
More Product Information