Cooled Incubators, KB
Supplier: VWR International
77938-554 77938-556 77938-558 77938-560 77938-562 77938-540
77938-554EA 14648.78 CAD
77938-554 77938-556 77938-558 77938-560 77938-562 77938-540
Cooled Incubators, KB
Incubators Cooled Incubators
The powerful KB Series of cooling incubators for microorganisms are able to incubate with exceptional precision even when fully loaded. Units have a stainless-steel inner chamber, feature proven APT.line™ preheating chamber technology, have a temperature range of 0 to 70 °C, hot-air disinfection routine, and can be programmed for weekend operation with ease. This KB unit consumes as much as 40% less energy than the previous model.
  • Energy-efficient cooling with modern inverter cooling unit
  • Cooling with climate-neutral R-600a refrigerant (GWP is 3)
  • Door
More Product Information