Constant Climate Chambers, KBF PRO
Supplier: Binder
9020-0439 9020-0440 9020-0441 9020-0442 9020-0443 9020-0444
77938-528EA 41708.84 CAD
77938-528 77938-530 77938-532 77938-534 77938-536 77938-538
Constant Climate Chambers, KBF PRO
Testing Chambers Climatic Chambers
A powerful all-rounder among constant climate chambers: Equipped with all kinds of sophisticated technology, the KBF PRO with proven APT.line™ preheating chamber technology takes any material test in its stride - whether it takes place at –20 °C or under THB conditions at 85 °C and 85% relative humidity.
  • Energy-efficient cooling with modern inverter cooling unit
  • Cooling with climate-neutral R-290 refrigerant (GWP is 3)
  • Door heating to prevent condensation
  • Adjustable fan speed
  • Humidity regulation with capacitive humidity sensor and responsive steam
More Product Information