Fite Control Slide

Supplier: StatLab
CS-FITE/25
77907-904PK 314.51 CAD
77907-904
Fite Control Slide
Slides Microscopy Control Slides
Fite control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.

  • Includes one stained slide
  • Tissue: known positive
  • Special stain or antibody: Fite's stain

Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
New Product
Order Now

SPECIFICATIONS

Species
Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR