DuPont™ Tychem® 6000 SFR Coveralls
Supplier: DuPont
FS127TLT2X000600 FS127TLT3X000600 FS127TLT4X000600 FS127TLT5X000600 FS127TLTLG000600 FS127TLTMD000600 FS127TLTSM000600 FS127TLTXL000600
77889-276CS 931.81 CAD
77889-276 77889-274 77889-280 77889-268 77889-278 77889-270 77889-282 77889-272
DuPont™ Tychem® 6000 SFR Coveralls
Disposable Garments
DuPont™ Tychem® 6000 SFR coveralls feature a respirator fit hood, front zipper closure, chin flap, double storm flap, elastic face opening, elastic wrists, elastic ankles, elastic waist, taped seams, and pin lock zipper.
  • Respirator-fit hood
  • Covered elastic face opening
  • Covered elastic openings at wrists and ankles for tighter fit
  • Elastic at sides of waist for better fit
  • Chin flap with double-sided adhesive tape
  • Double storm flap covers the zipper, which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion
  • Taped seams provide
More Product Information