β-Glc-PEG3-Alkyne 95%

Supplier: AFG Bioscience

C52004-25MG C52004-50MG C52004-100MG C52004-10MG
76619-728EA 473.48 CAD
76619-728 76619-730 77888-829 77888-830
β-Glc-PEG3-Alkyne 95%
β-Glc-PEG3-Alkyne
Formula: C₁₅H₂₆O₉
Storage Temperature: Freezer
CAS Number: 1072903-76-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR