Unit Dose Sample Die for Model I, IV, and V Powder and Liquid Samplers
Supplier: GLOBEPHARMA
M1.4.5.6.CE.SS UD.3.3.SD.SS UD.3.4.SD.SS UD.3.5.SD.SS UD.3.6.SD.SS UD.3.7.SD.SS UD.3.8.SD.SS UD.0.3.SD.SS UD.0.4.SD.SS UD.0.5.SD.SS UD.0.6.SD.SS UD.0.7.SD.SS UD.0.8.SD.SS UD.0.9.SD.SS UD.1.0.SD.SS UD.1.1.SD.SS UD.1.2.SD.SS UD.1.3.SD.SS UD.1.4.SD.SS UD.1.5.SD.SS UD.1.6.SD.SS UD.1.7.SD.SS UD.1.8.SD.SS UD.1.9.SD.SS UD.2.0.SD.SS UD.2.1.SD.SS UD.2.2.SD.SS UDSP.2.9.SD.SS UDSP.3.0.SD.SS UD.0.1.SD.SS UD.0.2.SD.SS UD.2.3.SD.SS UD.2.4.SD.SS UD.2.5.SD.SS UD.2.6.SD.SS UD.2.7.SD.SS UD.2.8.SD.SS UD.2.9.SD.SS UD.3.0.SD.SS UD.3.1.SD.SS UD.3.2.SD.SS UDSP.0.9.SD.SS UDSP.1.0.SD.SS UDSP.1.1.SD.SS UDSP.1.2.SD.SS UDSP.1.3.SD.SS UDSP.1.4.SD.SS UDSP.1.5.SD.SS UDSP.1.6.SD.SS UDSP.1.7.SD.SS UDSP.1.8.SD.SS UDSP.1.9.SD.SS UDSP.2.0.SD.SS UDSP.2.1.SD.SS UDSP.2.2.SD.SS UDSP.2.3.SD.SS UDSP.2.4.SD.SS UDSP.2.5.SD.SS UDSP.2.6.SD.SS UDSP.2.7.SD.SS UDSP.2.8.SD.SS UD.HB.SS UD.SB.SS UDSP.0.1.SD.SS UDSP.0.2.SD.SS UDSP.0.3.SD.SS UDSP.0.4.SD.SS UDSP.0.5.SD.SS UDSP.0.6.SD.SS UDSP.0.7.SD.SS UDSP.0.8.SD.SS UD.3.9.SD.SS UD.4.0.SD.SS
77887-288EA 198.76 CAD
77887-288 77887-450 77887-706 77887-408 77887-126 77887-280 77887-426 77887-578 77888-406 77887-154 77888-536 77888-074 77887-160 77887-616 77887-784 77887-082 77887-486 77888-414 77888-398 77887-674 77887-974 77887-258 77887-330 77888-510 77887-914 77888-474 77887-104 77887-524 77887-354 77887-862 77888-528 77888-012 77888-488 77887-840 77888-064 77888-082 77887-420 77887-470 77888-172 77888-454 77888-066 77888-434 77887-672 77887-440 77888-446 77887-966 77887-530 77888-182 77887-904 77887-636 77887-710 77887-602 77887-610 77888-472 77888-524 77887-212 77887-220 77887-326 77887-442 77887-352 77888-490 77887-214 77888-026 77887-558 77887-630 77888-158 77887-014 77887-994 77888-104 77887-936 77887-006 77887-858 77888-058
Unit Dose Sample Die for Model I, IV, and V Powder and Liquid Samplers
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: Made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: Engineered for quick disassembly and
More Product Information