Powder and Liquid Samplers, Model IC
Supplier: GLOBEPHARMA
M1.C.0.1.SD.SS M1.C.0.2.SD.SS M1.C.0.3.SD.SS M1.C.0.8.SD.SS M1.C.0.9.SD.SS M1.C.1.0.SD.SS M1.C.1.1.SD.SS M1.C.1.2.SD.SS M1.C.3.5.SD.SS M1.C.3.6.SD.SS M1.C.3.7.SD.SS M1.C.3.8.SD.SS M1.C.3.9.SD.SS M1.C.4.0.SD.SS M1.C.6.SS M1.C.2.5.SD.SS M1.C.2.6.SD.SS M1.C.2.7.SD.SS M1.C.2.8.SD.SS M1.C.2.9.SD.SS M1.C.3.0.SD.SS M1.C.3.1.SD.SS M1.C.3.2.SD.SS M1.C.3.3.SD.SS M1.C.3.4.SD.SS M1.C.0.4.SD.SS M1.C.0.5.SD.SS M1.C.0.6.SD.SS M1.C.1.7.SD.SS M1.C.1.8.SD.SS M1.C.1.9.SD.SS M1.C.12.SS M1.C.18.SS M1.C.2.0.SD.SS M1.C.2.1.SD.SS M1.C.2.2.SD.SS M1.C.2.3.SD.SS M1.C.2.4.SD.SS M1.C.1.3.SD.SS M1.C.1.4.SD.SS M1.C.1.5.SD.SS M1.C.1.6.SD.SS
77888-076EA 323 CAD
77888-076 77888-368 77887-262 77887-682 77887-766 77887-064 77887-864 77887-806 77888-420 77888-462 77887-218 77887-360 77887-956 77888-092 77888-036 77888-174 77887-500 77887-308 77887-576 77888-494 77888-354 77887-640 77887-302 77888-122 77887-540 77887-920 77887-830 77888-046 77887-866 77887-052 77887-138 77887-594 77888-038 77888-060 77888-068 77887-036 77887-646 77887-804 77888-480 77887-978 77887-180 77887-760
Powder and Liquid Samplers, Model IC
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: Made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: Engineered for quick disassembly and
More Product Information