Powder and Liquid Samplers, ID
Supplier: GLOBEPHARMA
M1.D.0.4.SD.SS M1.D.0.5.SD.SS M1.D.0.6.SD.SS M1.D.0.7.SD.SS M1.D.0.8.SD.SS M1.D.0.9.SD.SS M1.D.1.0.SD.SS M1.D.1.1.SD.SS M1.D.1.1.SS M1.D.1.2.SD.SS M1.D.3.7.SD.SS M1.D.3.8.SD.SS M1.D.3.9.SD.SS M1.D.4.0.SD.SS M1.D.4.1.SD.SS M1.D.4.1.SS M1.D.4.2.SD.SS M1.D.4.3.SD.SS M1.D.4.4.SD.SS M1.D.4.5.SD.SS M1.D.9.1.SD.SS M1.D.9.2.SD.SS M1.D.2.1.SD.SS M1.D.2.1.SS M1.D.2.10.SS M1.D.2.2.SD.SS M1.D.2.3.SD.SS M1.D.2.4.SD.SS M1.D.2.5.SD.SS M1.D.2.6.SD.SS M1.D.2.7.SD.SS M1.D.2.8.SD.SS M1.D.6.12.SS M1.D.6.2.SD.SS M1.D.6.3.SD.SS M1.D.6.4.SD.SS M1.D.6.5.SD.SS M1.D.6.6.SD.SS M1.D.6.7.SD.SS M1.D.6.8.SD.SS M1.D.6.9.SD.SS M1.D.7.0.SD.SS M1.D.0.1.SD.SS M1.D.0.2.SD.SS M1.D.0.3.SD.SS M1.D.7.1.SD.SS M1.D.7.2.SD.SS M1.D.7.3.SD.SS M1.D.7.4.SD.SS M1.D.7.5.SD.SS M1.D.7.6.SD.SS M1.D.7.7.SD.SS M1.D.7.8.SD.SS M1.D.7.9.SD.SS M1.D.8.0.SD.SS M1.D.4.6.SD.SS M1.D.4.7.SD.SS M1.D.4.8.SD.SS M1.D.4.8.SS M1.D.4.9.SD.SS M1.D.5.0.SD.SS M1.D.5.1.SD.SS M1.D.5.1.SS M1.D.5.10.SS M1.D.5.2.SD.SS M1.D.5.3.SD.SS M1.D.5.4.SD.SS M1.D.5.5.SD.SS M1.D.5.6.SD.SS M1.D.5.7.SD.SS M1.D.5.8.SD.SS M1.D.5.9.SD.SS M1.D.6.0.SD.SS M1.D.6.1.SD.SS M1.D.6.1.SS M1.D.1.3.SD.SS M1.D.1.4.SD.SS M1.D.1.5.SD.SS M1.D.1.5.SS M1.D.1.6.SD.SS M1.D.1.7.SD.SS M1.D.1.8.SD.SS M1.D.1.9.SD.SS M1.D.10.SD.SS M1.D.2.0.SD.SS M1.D.8.1.SD.SS M1.D.8.2.SD.SS M1.D.8.3.SD.SS M1.D.8.4.SD.SS M1.D.8.5.SD.SS M1.D.8.6.SD.SS M1.D.8.7.SD.SS M1.D.8.8.SD.SS M1.D.8.9.SD.SS M1.D.9.0.SD.SS M1.D.2.9.SD.SS M1.D.3.0.SD.SS M1.D.3.1.SD.SS M1.D.3.1.SS M1.D.3.2.SD.SS M1.D.3.3.SD.SS M1.D.3.4.SD.SS M1.D.3.5.SD.SS M1.D.3.6.SD.SS M1.D.3.6.SS M1.D.9.3.SD.SS M1.D.9.4.SD.SS M1.D.9.5.SD.SS M1.D.9.6.SD.SS M1.D.9.7.SD.SS M1.D.9.8.SD.SS M1.D.9.9.SD.SS M1.D.BD.SD
77887-820EA 372.69 CAD
77887-820 77887-010 77887-210 77888-098 77887-454 77888-194 77887-574 77888-008 77887-562 77887-626 77888-110 77887-430 77887-614 77887-536 77887-034 77887-644 77887-130 77887-414 77887-642 77887-664 77887-204 77887-514 77887-108 77887-238 77887-522 77887-110 77887-520 77887-474 77887-714 77888-072 77887-538 77887-194 77887-824 77887-284 77887-338 77887-398 77888-526 77887-570 77888-384 77887-754 77888-160 77888-056 77887-070 77888-216 77887-740 77887-718 77887-230 77887-582 77888-352 77888-022 77887-794 77887-378 77888-078 77887-396 77887-458 77887-924 77887-846 77888-440 77888-484 77887-114 77887-100 77887-168 77887-876 77887-588 77887-676 77888-448 77887-948 77887-742 77887-808 77887-428 77887-268 77887-448 77887-878 77887-662 77887-246 77887-254 77887-422 77887-512 77887-814 77888-402 77888-116 77888-032 77887-658 77888-356 77887-762 77888-220 77888-154 77887-836 77888-476 77887-844 77888-184 77887-444 77887-182 77887-882 77887-478 77887-496 77887-232 77887-950 77887-898 77888-146 77887-118 77887-120 77887-062 77887-768 77887-142 77887-390 77887-860 77887-178 77887-004 77887-260 77888-378 77888-458 77887-290
Powder and Liquid Samplers, ID
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: engineered for quick disassembly and
More Product Information