Powder and Liquid Samplers, XVIB
Supplier: GLOBEPHARMA
M16B.SD.0.4CC M16B.SD.0.5CC M16B.SD.0.6CC M16B.SD.0.7CC M16B.SD.0.8CC M16B.SD.0.9CC M16B.SD.1.0CC M16B.SD.1.1CC M16B.SD.1.2CC M16B.SD.1.3CC M16B.TSD.0.4CC M16B.TSD.0.5CC M16B.TSD.0.6CC M16B.TSD.0.7CC M16B.TSD.0.8CC M16B.TSD.0.9CC M16B.TSD.1.0CC M16B.TSD.1.1CC M16B.TSD.1.2CC M16B.TSD.1.3CC M16B.TSD.3.4CC M16B.TSD.3.5CC M16B.TSD.3.6CC M16B.TSD.3.7CC M16B.TSD.3.8CC M16B.TSD.3.9CC M16B.TSD.4.0CC M16B.TSD.4.1CC M16B.TSD.4.2CC M16B.TSD.4.3CC M16B.SD.3.4CC M16B.SD.3.5CC M16B.SD.3.6CC M16B.SD.3.7CC M16B.SD.3.8CC M16B.SD.3.9CC M16B.SD.4.0CC M16B.SD.4.1CC M16B.SD.4.2CC M16B.SD.4.3CC M16B.3.S.SS M16B.SD.2.4CC M16B.SD.2.5CC M16B.SD.2.6CC M16B.SD.2.7CC M16B.SD.2.8CC M16B.SD.2.9CC M16B.SD.3.0CC M16B.SD.3.1CC M16B.SD.3.2CC M16B.SD.3.3CC M16B.TSD.1.4CC M16B.TSD.1.5CC M16B.TSD.1.6CC M16B.TSD.1.7CC M16B.TSD.1.8CC M16B.TSD.1.9CC M16B.TSD.2.0CC M16B.TSD.2.1CC M16B.TSD.2.2CC M16B.TSD.2.3CC M16B.TSD.4.4CC M16B.TSD.4.5CC M16B.TSD.4.6CC M16B.TSD.4.7CC M16B.TSD.4.8CC M16B.TSD.4.9CC M16B.TSD.5.0CC M16BT.3.S.SS M16BT.6.S.SS M16BT.EP.SET M16B.SD.1.4CC M16B.SD.1.5CC M16B.SD.1.6CC M16B.SD.1.7CC M16B.SD.1.8CC M16B.SD.1.9CC M16B.SD.2.0CC M16B.SD.2.1CC M16B.SD.2.2CC M16B.SD.2.3CC M16B.SD.4.4CC M16B.SD.4.5CC M16B.SD.4.6CC M16B.SD.4.7CC M16B.SD.4.8CC M16B.SD.4.9CC M16B.SD.5.0CC M16B.TSD.0.1CC M16B.TSD.0.2CC M16B.TSD.0.3CC M16B.TSD.2.4CC M16B.TSD.2.5CC M16B.TSD.2.6CC M16B.TSD.2.7CC M16B.TSD.2.8CC M16B.TSD.2.9CC M16B.TSD.3.0CC M16B.TSD.3.1CC M16B.TSD.3.2CC M16B.TSD.3.3CC M16B.6.S.SS M16B.EP.SET M16B.MP.12 M16B.MP.14 M16B.MP.316 M16B.MP.38 M16B.SD.0.1CC M16B.SD.0.2CC M16B.SD.0.3CC
77888-478EA 1031.09 CAD
77888-478 77887-184 77888-442 77888-084 77887-000 77887-654 77887-792 77887-980 77888-504 77887-490 77888-530 77887-402 77887-226 77887-802 77887-544 77887-196 77888-432 77887-548 77887-652 77888-176 77887-842 77887-854 77887-376 77888-364 77888-452 77887-068 77887-208 77887-056 77888-532 77888-062 77887-198 77887-482 77888-134 77888-520 77887-012 77887-566 77888-180 77887-510 77888-112 77887-494 77887-838 77887-982 77887-200 77887-424 77887-296 77887-902 77887-022 77888-498 77888-044 77887-298 77888-450 77887-952 77887-132 77888-050 77887-250 77888-382 77888-142 77887-028 77887-090 77887-412 77887-600 77887-564 77888-150 77887-492 77887-382 77887-782 77887-048 77887-868 77888-362 77887-074 77888-014 77888-006 77887-084 77887-690 77887-638 77887-518 77888-124 77887-526 77888-106 77888-100 77887-372 77887-992 77887-124 77887-032 77888-102 77888-000 77888-186 77887-096 77887-532 77887-092 77887-172 77888-522 77887-938 77887-316 77888-138 77888-380 77887-344 77888-512 77887-890 77887-790 77887-774 77887-998 77888-168 77887-294 77887-388 77887-128 77887-094 77887-632 77888-412 77887-248
Powder and Liquid Samplers, XVIB
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: Made from high-quality 316L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: Engineered for quick disassembly and
More Product Information