Duplicate Sample Powder and Liquid Samplers, IV
Supplier: GLOBEPHARMA
M4.6.12.SS M4.2.1.SS M4.2.5.SS M4.3.1.SS M4.3.7.SS M4.4.1.SS M4.4.8.SS M4.5.1.SS M4.5.10.SS M4.6.1.SS
77887-060EA 7441.23 CAD
77887-060 77888-052 77887-650 77887-026 77888-428 77888-416 77887-066 77887-884 77888-464 77888-118
Duplicate Sample Powder and Liquid Samplers, IV
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations. Duplicate sample devices are constructed so that each port holds two dies.
  • Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: Made from high-quality 316L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease
More Product Information