Extended Length LfTS fit Pipette Tip Reload Systems, Non Sterile
Supplier: VistaLab
4080-1042
77873-988CS 105.86 CAD
77873-988
Extended Length LfTS fit Pipette Tip Reload Systems, Non Sterile
Pipette Tips
Designed to provide a low-force tip solution for more comfortable pipetting; less force needed to acquire tips.
  • LfTS reload systems offer a convenient and economical way to refill empty tip racks, saving both time and money
  • Reload systems dramatically reduce plastic waste by reusing empty racks
  • Fine tip for improved accuracy and precision
  • Polypropylene
These 10 μl extended length tips have a maximum volume of 20 μl and are compatible with 2, 10, and 20 μl pipettes. LfTS reload system tips, wafers, and racks are fully autoclavable.