Corepoint Scientific™ Temperature Test Chambers 72 Cu. Ft., Horizon Scientific
Supplier: Horizon Scientific
NSRI723WSG/0 NSRI723WSW/0
77873-416EA 41250.64 CAD
77873-416 77873-420
Corepoint Scientific™ Temperature Test Chambers 72 Cu. Ft., Horizon Scientific
Testing Chambers Temperature Test Chambers
Transform your product testing with Corpoint® Scientific Temperature Test Chambers.
  • Programmable PID controller
  • Variable speed compressor
  • Broad temperature range
  • USB data port
  • Natural hydrocarbon refrigerants
Ensure the quality and durability of your products with Corepoint® Scientific Temperature Test Chambers. These chambers provide a controlled temperature environment to deliver superior product testing backed by FDA and ICH standards. Improve product quality, test packaging, increase shelf life, and enhance safety with these chambers. With naturalMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Int. W×D×H 1695×594×1511 mm (66.75×23.375×59.5")
Ext. W×D×H 2057×921×2070 mm (81×36.25×81.5")