Saturix Presaturated Cleanroom Wipes, 96% Presaturated Sachets
Supplier: FG CLEAN WIPES TE
6-C43 6-C86-EFP
77871-044CS 467.68 CAD
77871-044 77871-046
Saturix Presaturated Cleanroom Wipes, 96% Presaturated Sachets
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Convenience, consistency, and cleanliness.
  • 96% IPA, 4% DI water
  • Single-use, easy tear foil packages
  • Clean, pure, durable, and ultra-low linting non-woven polyester
  • Ease of use for workstations and on-the-go field support
  • Industry recommended for the electrofusion process
  • Fast and efficient cleaning, dries in seconds
  • Consistency in solvent application
  • Safer to ship, store, and use
These wipes feature a high concentration of 96% IPA on pure polyester in a single-use, easy tear foil package or sachet. Designed for tough wet wiping in the field. The ultra-
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable