AlphaTec® ActivArmr® R011BUL Ultra-Lightweight Electrical Insulated Gloves, Class 0, Ansell
Supplier: Ansell Healthcare
RIG011BUL090 RIG011BUL100 RIG011BUL110 RIG011BUL070 RIG011BUL080
77867-868CS 1045.23 CAD
77867-868 77867-870 77867-872 77867-864 77867-866
AlphaTec® ActivArmr® R011BUL Ultra-Lightweight Electrical Insulated Gloves, Class 0, Ansell
Gloves
Up to 20% thinner glove design offers electrical protection of up to 1000 V AC/1500V DC.
  • Form fitting design is up to 20% thinner, resulting in improved dexterity and tactility
  • All round protection against electrical risks (up to 1000 V AC/1500V DC) and Arc flashes
  • Silicone-free and Prop 65 compliant, and In-house testing of EN388
Up to 20% thinner than comparable Class 0 gloves, the new form fitting design provides unmatched dexterity and tactility. These gloves meet requirements for Class 0 (1000 V AC/1500 V DC) protection. They are Arc flash-
More Product Information