Edge® 48-969 Medium-Duty Cut Protection Industrial Gloves, Ansell
Supplier: Ansell Healthcare
48969090 48969100 48969110 48969060 48969070 48969080
77867-848CS 1750.17 CAD
77867-848 77867-850 77867-852 77867-842 77867-844 77867-846
Edge® 48-969 Medium-Duty Cut Protection Industrial Gloves, Ansell
Gloves
Economical solution for medium-duty applications requiring very high cut resistance, grip, and oil-repelling performance in oily environments.
  • NBR coated finish offers high grip in oily environments
  • Bright yellow design ensures high visibility
  • Thermal resistance allows the ability to confidently handle hot components
  • Superior cut-resistant lining for impact gloves with 360° cut resistance
  • Patented TPR design provides knuckle, thumb, and finger-length impact protection
Featuring a fully NBR-coated seamless liner with a rough palm, Edge® 48-969
More Product Information