Anti-KRT6A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A27208-100UG A27208-50UG
77741-894EA 521.4 CAD
77741-894 77858-759
Anti-KRT6A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: KRT6A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol KRT6A
Environmentally Preferable
Antigen synonyms CK-6A|CK-6D|CK6A|CK6C|CK6D|Cytokeratin-6A|Cytokeratin-6D|K2C6A_HUMAN|K6A|K6C|K6D|keratin 6A|Keratin|type II cytoskeletal 6A|Keratin-6A|KRT6C|KRT6D|Type-II keratin Kb6
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P02538
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Keratin, type II cytoskeletal 6A protein (2-564AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR