Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25010-50UL
77857-225EA 355.5 CAD
77857-225
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, ICC: 1:10 to 1:100, IF: 1:10 to 1:100, IP: 1:200 to 1:2000

Type:
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR