Anti-DNPEP Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A19748-50UG
77854-304EA 355.5 CAD
77854-304
Anti-DNPEP Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: DNPEP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol DNPEP
Environmentally Preferable
Antigen synonyms Aspartyl aminopeptidase|ASPEP|DAP|DNPEP_HUMAN
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9ULA0
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Aspartyl aminopeptidase protein (279-386AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR