Anti-IgG Fab Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A17171-500UG
77853-035EA 395 CAD
77853-035
Anti-IgG Fab Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG Fab
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol IgG Fab
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
ELISA Yes
Immunogen Human IgG Fab fragment
Form Liquid
Purification Caprylic acid and ammonium sulfate precipitation
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR