Anti-CEACAM5 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A16995-50UL
77852-946EA 355.5 CAD
77852-946
Anti-CEACAM5 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200

Type:
Antigen: CEACAM5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol CEACAM5
Environmentally Preferable
Antigen synonyms Carcinoembryonic antigen|Carcinoembryonic antigen-related cell adhesion molecule 5|CD66e|CEA|CEAM5_HUMAN|DKFZp781M2392|Meconium antigen 100|OTTHUMP00000199032|OTTHUMP00000199033|OTTHUMP00000199034
Clonality Recombinant Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID P06731
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen Recombinant human Carcinoembryonic antigen-related cell adhesion molecule 5 protein (1-255AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR