Anti-MET Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73880-100UL
77848-691EA 521.4 CAD
77848-691
Anti-MET Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:3000

Type:
Antigen: MET
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR