Anti-DSC1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73803-100UL A73803-50UL
77848-561EA 521.4 CAD
77848-561 77848-562
Anti-DSC1 Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: DSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR