Anti-AXIN2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A72471-100UL A72471-50UL
77846-318EA 521.4 CAD
77846-318 77846-319
Anti-AXIN2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: AXIN2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol AXIN2
Environmentally Preferable
Antigen synonyms Axil|Axin like protein|Axin-2|Axin-like protein|Axin2|AXIN2_HUMAN|Axis inhibition protein 2|Conductin|DKFZp781B0869|MGC10366|MGC126582
Clonality Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9Y2T1
Isotype IgG
ELISA Yes
Immunogen Peptide sequence from Human Axin-2 protein (56-73AA)
Form Liquid
Purification Antigen affinity purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR