Anti-DSE Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A71273-100UG A71273-50UG
77844-068EA 521.4 CAD
77844-068 77844-069
Anti-DSE Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: DSE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol DSE
Environmentally Preferable
Antigen synonyms Chondroitin glucuronate 5 epimerase|Chondroitin-glucuronate 5-epimerase|Dermatan sulfate epimerase|Dermatan-sulfate epimerase|DS epimerase|DSE_HUMAN|DSEPI|OTTHUMP00000040406|SART 2|SART-2|Squamous cell carcinoma antigen recognized by T cells 2|Squamous cell carcinoma antigen recognized by T-cells 2
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID Q9UL01
Isotype IgG
ELISA Yes
Immunogen Recombinant human Dermatan-sulfate epimerase protein (23-270AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR