Anti-ompA Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A57158-100UG A57158-50UG
77827-599EA 521.4 CAD
77827-599 77827-600
Anti-ompA Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: ompA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rickettsia conorii
New Product
Order Now

SPECIFICATIONS

Antigen symbol ompA;RC1273;Outer membrane protein A;190 kDa antigen;Cell surface antigen;rOmp A;rOmpA) [Cleaved into: 120 kDa surface-exposed protein;120 kDa outer membrane protein OmpA);32 kDa beta peptide]
Environmentally Preferable
Antigen name Recombinant Rickettsia conorii Outer membrane protein A protein (1734-2021AA)
Clonality Recombinant Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Rickettsia conorii
Host Rabbit
Gene ID Q52657
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR