Anti-PTRH2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A57009-100UL A57009-50UL
77827-326EA 521.4 CAD
77827-326 77827-327
Anti-PTRH2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:10 to 1:50

Type:
Antigen: PTRH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR